CO1H324AAE94 CombiPhos Catalysts Inc

4-bromo-1-(3-fluorophenyl)-1H-imidazol-2-amine

  • Molecular Weight : 256.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart