CO1H324AAEBA CombiPhos Catalysts Inc

1-ethyl-2-methoxy-1H-imidazol-4-amine

  • Cas Number : 1891061-31-6
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart