CO1H324AAEE5 CombiPhos Catalysts Inc

4-bromo-3-iodopyridin-2-ol

  • Molecular Weight : 299.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart