CO1H324AAFAB CombiPhos Catalysts Inc

5-bromo-2-methoxy-1-(2-(trifluoromethyl)phenyl)-1H-imidazole

  • Molecular Weight : 321.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart