CO1H324AAFB0 CombiPhos Catalysts Inc

2,5-dibromo-1-(4-(trifluoromethyl)phenyl)-1H-imidazole

  • Molecular Weight : 369.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart