CO1H324AAFE9 CombiPhos Catalysts Inc

5-bromo-1H-imidazol-2-amine

  • Cas Number : 1211524-33-2
  • Molecular Weight : 161.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart