CO1H324AB095 CombiPhos Catalysts Inc

5-bromo-1-(o-tolyl)-1,3-dihydro-2H-imidazol-2-one

  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart