CO1H324AB0FC CombiPhos Catalysts Inc

tert-butyl 3-(6-(tert-butoxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)azetidine-1-carboxylate

  • Cas Number : 2256754-82-0
  • Molecular Weight : 432.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart