CO1H324AB13B CombiPhos Catalysts Inc

2-(3-fluoro-4-(tetrahydrofuran-3-yl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 2256755-38-9
  • Molecular Weight : 292.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart