CO1H324AB146 CombiPhos Catalysts Inc

2-fluoro-6-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)pyridine

  • Cas Number : 2256755-47-0
  • Molecular Weight : 289.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart