CO1H324AB196 CombiPhos Catalysts Inc

1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,2,3-triazole

  • Cas Number : 2256756-21-3
  • Molecular Weight : 271.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart