CO1H324AB1B6 CombiPhos Catalysts Inc

2-chloro-4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,2,3-triazol-1-yl)pyridine

  • Cas Number : 2256758-02-6
  • Molecular Weight : 306.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart