CO1H324AB1B7 CombiPhos Catalysts Inc

2-methyl-6-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,2,3-triazol-1-yl)pyridine

  • Cas Number : 2256758-03-7
  • Molecular Weight : 286.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart