CO1H324AB261 CombiPhos Catalysts Inc

tert-butyl 3-(2-(dimethylamino)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazol-1-yl)pyrrolidine-1-carboxylate

  • Molecular Weight : 406.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart