CO1H324AB36E CombiPhos Catalysts Inc

3-Cyclopropylpiperidine

  • Molecular Weight : 125.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart