CO1H324AB387 CombiPhos Catalysts Inc

5-(tetrahydro-2H-pyran-2-yl)pyrazin-2-amine

  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart