CO1H324AB391 CombiPhos Catalysts Inc

4-chloro-2-(tetrahydro-2H-pyran-2-yl)pyrimidine

  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart