CO1H324AB3D9 CombiPhos Catalysts Inc

4-Bromo-1-(n-propyl)-2-(iso-propyl)imidazole

  • Molecular Weight : 231.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart