CO1H324AB3E5 CombiPhos Catalysts Inc

1-(Oxan-4-yl)-2-(1-methylethyl)-1H-imidazol-4-amine

  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart