CO1H324AB405 CombiPhos Catalysts Inc

4,5-Dibromo-1-cyclopropyl-2-ethyl-1H-imidazole

  • Molecular Weight : 293.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart