CO1H324AB41C CombiPhos Catalysts Inc

4,5-Dibromo-1-(N-Boc-piperidin-3-yl)-2-methyl-1H-imidazole

  • Molecular Weight : 423.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart