CO1H324AB433 CombiPhos Catalysts Inc

1-Cyclohexyl-1H-1,2,3-triazol-4-amine

  • Molecular Weight : 166.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart