CO1H324AB453 CombiPhos Catalysts Inc

4-Bromo-2-(oxan-3-yl)-1H-imidazole

  • Molecular Weight : 231.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart