CO1H324AB4F8 CombiPhos Catalysts Inc

4-Chloro-5-bromo-2-(tert-butyl)pyrimidine

  • Molecular Weight : 249.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart