CO1H324AB5CD CombiPhos Catalysts Inc

4-Chloro-2-hydroxy-5-ethylpyrimidine

  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart