ENA018104150 Enamine

3-Nitro-4-piperidin-1-yl-benzoic acid

  • MDL number MFCD00157001
  • Cas Number : 26586-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00039

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00039
Availability details for
Quantity
  • Availability
  • Add to Cart