ENA018104153 Enamine

4-chloro-3-(thiophene-2-amido)benzoic acid

  • MDL number MFCD03147268
  • Cas Number : 379254-57-6
  • Molecular Weight : 281.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-167
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00042
Availability details for
Quantity
  • Availability
  • Add to Cart