ENA018104159 Enamine

3-[(2-chlorophenyl)sulfanyl]propanoic acid

  • MDL number MFCD02720703
  • Cas Number : 99585-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-93
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00048
Availability details for
Quantity
  • Availability
  • Add to Cart