ENA018104169 Enamine

2-[(4-bromo-2-methylphenyl)sulfanyl]acetic acid

  • MDL number MFCD00519331
  • Cas Number : 42943-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00059
Availability details for
Quantity
  • Availability
  • Add to Cart