ENA018104172 Enamine

4-{[(4-chlorophenyl)sulfanyl]methyl}benzoic acid

  • MDL number MFCD00494342
  • Cas Number : 88382-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00062

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00062
Availability details for
Quantity
  • Availability
  • Add to Cart