ENA018104174 Enamine

3-(4-Fluoro-phenyl)-2-thiophen-2-yl-acrylic acid

  • MDL number MFCD02725302
  • Cas Number : 104314-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185...187
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00065

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00065
Availability details for
Quantity
  • Availability
  • Add to Cart