ENA018104175 Enamine

4-[(2,6-dibromo-4-methylphenoxy)methyl]benzoic acid

  • MDL number MFCD03148788
  • Cas Number : 380170-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00066
Availability details for
Quantity
  • Availability
  • Add to Cart