ENA018104201 Enamine

4-Chloro-2-(4-fluoro-benzoylamino)-benzoic acid

  • MDL number MFCD02032354
  • Cas Number : 38464-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250...252
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00097

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00097
Availability details for
Quantity
  • Availability
  • Add to Cart