ENA018104205 Enamine

2-[(2-chlorophenyl)formamido]acetic acid

  • MDL number MFCD00457852
  • Cas Number : 16555-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00101
Availability details for
Quantity
  • Availability
  • Add to Cart