ENA018104208 Enamine

(4-Propionyl-phenoxy)-acetic acid

  • MDL number MFCD00455856
  • Cas Number : 6501-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00104

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00104
Availability details for
Quantity
  • Availability
  • Add to Cart