ENA018104219 Enamine

3-(3-Methoxy-phenyl)-2-phenyl-acrylic acid

  • MDL number MFCD00598898
  • Cas Number : 94501-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00115

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00115
Availability details for
Quantity
  • Availability
  • Add to Cart