ENA018104223 Enamine

3-(4-tert-butylphenoxy)propanoic acid

  • MDL number MFCD03147408
  • Cas Number : 23067-72-3
  • Molecular Weight : 222.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 52-53
Iupac Name 3-(4-tert-butylphenoxy)propanoic acid
Inchi Code 1S/C13H18O3/c1-13(2,3)10-4-6-11(7-5-10)16-9-8-12(14)15/h4-7H,8-9H2,1-3H3,(H,14,15)
Inchi Key MFWXGILWCBBIOC-UHFFFAOYSA-N

Description

Melting Point 52-53
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00120
Availability details for
Quantity
  • Availability
  • Add to Cart