ENA018104226 Enamine

3-(2,5-dimethylphenoxy)propanoic acid

  • MDL number MFCD03147282
  • Cas Number : 4673-48-7
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-119
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00123
Availability details for
Quantity
  • Availability
  • Add to Cart