ENA018104227 Enamine

3-(2-ethoxyphenoxy)propanoic acid

  • MDL number MFCD03147283
  • Cas Number : 379254-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00124
Availability details for
Quantity
  • Availability
  • Add to Cart