ENA018104243 Enamine

2-{1-[2-(morpholin-4-yl)-2-oxoethyl]cyclopentyl}acetic acid

  • MDL number MFCD03648408
  • Cas Number : 618392-72-6
  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-45
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00144
Availability details for
Quantity
  • Availability
  • Add to Cart