ENA018104249 Enamine

1-(4,4,4-Trifluoro-3-oxo-but-1-enyl)-pyrrolidine-2-carboxylic acid

  • MDL number MFCD01222392
  • Cas Number : 133992-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00152

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00152
Availability details for
Quantity
  • Availability
  • Add to Cart