ENA018104261 Enamine

2-(4,4,4-Trifluoro-3-oxo-but-1-enylamino)-benzoic acid

  • MDL number MFCD03150639
  • Cas Number : 380430-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00165

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00165
Availability details for
Quantity
  • Availability
  • Add to Cart