ENA018104270 Enamine

4-(2-cyanoacetamido)benzoic acid

  • MDL number MFCD03443334
  • Cas Number : 31557-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00174
Availability details for
Quantity
  • Availability
  • Add to Cart