ENA018104287 Enamine

3-(4-Nitro-1,3-dioxo-1,3-dihydro-isoindol-2-yl)-propionic acid

  • MDL number MFCD00523384
  • Cas Number : 15728-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00191

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00191
Availability details for
Quantity
  • Availability
  • Add to Cart