ENA018104290 Enamine

[Carboxymethyl-(4,4,4-trifluoro-3-oxo-but-1-enyl)-amino]-acetic acid

  • MDL number MFCD03965937
  • Cas Number : 565166-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00194

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00194
Availability details for
Quantity
  • Availability
  • Add to Cart