ENA018104304 Enamine

2-{[(3-chloro-4-methoxyphenyl)carbamoyl]methoxy}benzoic acid

  • MDL number MFCD03653055
  • Cas Number : 748776-43-4
  • Molecular Weight : 335.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00209
Availability details for
Quantity
  • Availability
  • Add to Cart