ENA018104306 Enamine

2-[({4-[(difluoromethyl)sulfanyl]phenyl}carbamoyl)methoxy]benzoic acid

  • MDL number MFCD03653053
  • Cas Number : 748776-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-193
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00211
Availability details for
Quantity
  • Availability
  • Add to Cart