ENA018104307 Enamine

2-{[(4-bromophenyl)carbamoyl]methoxy}benzoic acid

  • MDL number MFCD01459657
  • Cas Number : 329220-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-163
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00212
Availability details for
Quantity
  • Availability
  • Add to Cart