ENA018104314 Enamine

4-{[(4-bromophenyl)carbamoyl]methoxy}-3-methoxybenzoic acid

  • MDL number MFCD03651690
  • Cas Number : 522624-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00219
Availability details for
Quantity
  • Availability
  • Add to Cart