ENA018104320 Enamine

4-(4-ethoxyphenoxy)butanoic acid

  • MDL number MFCD03966913
  • Cas Number : 554405-58-2
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00225
Availability details for
Quantity
  • Availability
  • Add to Cart